Assay Details
Gene Symbol: VNN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015137426.2 | XP_014992912.2 | 662 | 146 | ||
XM_015137427.2 | XP_014992913.2 | 586 | 146 | |||
XM_015137428.2 | XP_014992914.2 | 686 | 146 |

More Information
Set Membership: |
![]() ![]() ![]() |
