Assay Details
Gene Symbol: FN3KRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194815.3 | NP_001181744.2 | 4 - 5 | 500 | 70 | |
GenBank mRNA | JU336402.1 | - | 4 - 5 | 517 | 70 | |
JU477220.1 | - | 4 - 5 | 485 | 70 | ||
JV048260.1 | - | 4 - 5 | 500 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
