Assay Details
Gene Symbol: DDX39A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193491.1 | NP_001180420.1 | 4 - 5 | 623 | 64 | |
XM_015123082.2 | XP_014978568.1 | 967 | 64 | |||
XM_015123083.2 | XP_014978569.1 | 1781 | 64 | |||
XM_015123084.1 | XP_014978570.1 | 766 | 64 | |||
XM_015123085.2 | XP_014978571.1 | 746 | 64 | |||
GenBank mRNA | JU327645.1 | - | 4 - 5 | 465 | 64 | |
JU473916.1 | - | 5 - 6 | 722 | 64 | ||
JV045867.1 | - | 5 - 6 | 686 | 64 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
