Assay Details
Gene Symbol: NDUFB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015129909.2 | XP_014985395.1 | 503 | 97 | ||
GenBank mRNA | JV507831.1 | - | 2 - 3 | 403 | 97 | |
JV715753.1 | - | 2 - 3 | 402 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
