Assay Details
Gene Symbol: FCSK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001107211.4 | XP_001107211.3 | 3378 | 83 | ||
XM_015126521.2 | XP_014982007.2 | 3407 | 83 | |||
XM_015126522.2 | XP_014982008.2 | 3508 | 83 | |||
XM_015126523.2 | XP_014982009.2 | 3335 | 83 | |||
XM_015126524.2 | XP_014982010.2 | 3445 | 83 | |||
XM_015126525.2 | XP_014982011.2 | 3282 | 83 | |||
XM_015126526.2 | XP_014982012.2 | 3053 | 83 | |||
XM_028841121.1 | XP_028696954.1 | 3772 | 83 | |||
XM_028841122.1 | XP_028696955.1 | 3336 | 83 | |||
XM_028841123.1 | XP_028696956.1 | 3210 | 83 | |||
XM_028841124.1 | XP_028696957.1 | 2725 | 83 | |||
GenBank mRNA | JU326106.1 | - | 22 - 23 | 3147 | 83 | |
JV046377.1 | - | 22 - 23 | 3173 | 83 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
