Assay Details
Gene Symbol: CFH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028851641.1 | XP_028707474.1 | 711 | 147 | ||
XM_028851648.1 | XP_028707481.1 | 711 | 147 | |||
XM_028851652.1 | XP_028707485.1 | 711 | 147 | |||
XM_028851653.1 | XP_028707486.1 | 711 | 147 | |||
XM_028851659.1 | XP_028707492.1 | 711 | 147 | |||
XM_028851660.1 | XP_028707493.1 | 711 | 147 | |||
GenBank mRNA | JU337428.1 | - | 443 | 147 | ||
JV453348.1 | - | 418 | 147 | |||
JV489867.1 | - | 426 | 147 | |||
JX277092.1 | - | 423 | 147 | |||
JX277093.1 | - | 423 | 147 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
