Assay Details
Gene Symbol: BUD23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266959.1 | NP_001253888.1 | 392 | 82 | ||
XM_028845071.1 | XP_028700904.1 | 518 | 82 | |||
GenBank mRNA | JU332790.1 | - | 398 | 82 | ||
JU332791.1 | - | 398 | 82 | |||
JU332792.1 | - | 398 | 82 | |||
JU474210.1 | - | 398 | 82 | |||
JV043191.1 | - | 392 | 82 | |||
JV706194.1 | - | 414 | 82 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
