Assay Details
Gene Symbol: LOC716834 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001114328.4 | XP_001114328.2 | 3240 | 87 | ||
XM_015151038.2 | XP_015006524.2 | 3203 | 87 | |||
GenBank mRNA | JU323357.1 | - | 23 - 24 | 87 | ||
JU471944.1 | - | 23 - 24 | 87 | |||
JV044860.1 | - | 23 - 24 | 87 | |||
JV452949.1 | - | 23 - 24 | 2973 | 87 | ||
JV635650.1 | - | 23 - 24 | 87 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
