Assay Details
Gene Symbol: MANF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015130831.1 | XP_014986317.1 | 374 | 94 | ||
GenBank mRNA | JU327052.1 | - | 2 - 3 | 216 | 94 | |
JV519846.1 | - | 2 - 3 | 186 | 94 | ||
JV593230.1 | - | 2 - 3 | 262 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
