Assay Details
Gene Symbol: PRXL2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002805660.3 | XP_002805706.2 | 579 | 87 | ||
XM_015147134.2 | XP_015002620.1 | 596 | 87 | |||
XM_015147135.2 | XP_015002621.1 | 631 | 87 | |||
XM_015147136.2 | XP_015002622.1 | 493 | 87 | |||
XM_028826411.1 | XP_028682244.1 | 487 | 87 | |||
XM_028826412.1 | XP_028682245.1 | 424 | 87 | |||
GenBank mRNA | JU473710.1 | - | 4 - 5 | 539 | 87 | |
JU473711.1 | - | 4 - 5 | 539 | 87 | ||
JV045354.1 | - | 4 - 5 | 538 | 87 | ||
JV635801.1 | - | 4 - 5 | 523 | 87 | ||
JV668307.1 | - | 4 - 5 | 523 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
