Assay Details
Gene Symbol: RPS16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265778.2 | NP_001252707.1 | 1 | 60 | 81 | |
GenBank mRNA | JV590103.1 | - | 1 | 61 | 81 | |
JV667450.1 | - | 1 | 63 | 81 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001265778.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
