Assay Details
Gene Symbol: DUSP21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265675.1 | NP_001252604.1 | 1 | 118 | 179 | |
GenBank mRNA | JV636313.1 | - | 1 | 118 | 179 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001265675.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
