Assay Details
Gene Symbol: MYCT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194146.1 | NP_001181075.1 | 1 - 2 | 195 | 109 | |
GenBank mRNA | JU329577.1 | - | 1 - 2 | 171 | 109 | |
JV723967.1 | - | 1 - 2 | 96 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
