Assay Details
Gene Symbol: FKBP1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266723.1 | NP_001253652.1 | 2 - 3 | 225 | 136 | |
XM_015111806.2 | XP_014967292.1 | 270 | 136 | |||
GenBank mRNA | JU320307.1 | - | 2 - 3 | 225 | 136 | |
JU321344.1 | - | 2 - 3 | 225 | 136 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
