Assay Details
Gene Symbol: SENP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193847.2 | NP_001180776.2 | 1 | 1311 | 110 | |
XM_015142594.2 | XP_014998080.2 | 1510 | 110 | |||
XM_015142595.2 | XP_014998081.2 | 1496 | 110 | |||
XM_015142596.2 | XP_014998082.2 | 1376 | 110 | |||
GenBank mRNA | JU325827.1 | - | 1 | 947 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
