Assay Details
Gene Symbol: KBTBD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015115578.2 | XP_014971064.2 | 432 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015115578.2 transcripts.
Set Membership: |
![]() ![]() |
