Assay Details
Gene Symbol: DDB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260837.1 | NP_001247766.1 | 4 - 5 | 778 | 84 | |
GenBank mRNA | JU472367.1 | - | 4 - 5 | 778 | 84 |
Gene Symbol: LOC114672264 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028833470.1 | XP_028689303.1 | 312 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
