Assay Details
Gene Symbol: MPZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001321055.2 | NP_001307984.1 | 5 - 6 | 711 | 116 | |
NM_001321056.2 | NP_001307985.1 | 5 - 6 | 711 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
