Assay Details
Gene Symbol: COP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266265.1 | NP_001253194.1 | 8 - 9 | 1126 | 74 | |
XM_015123338.2 | XP_014978824.1 | 1459 | 74 | |||
XM_015123346.2 | XP_014978832.1 | 1411 | 74 | |||
XM_015123351.2 | XP_014978837.1 | 1399 | 74 | |||
XM_015123363.2 | XP_014978849.1 | 1274 | 74 | |||
GenBank mRNA | JU327299.1 | - | 5 - 6 | 364 | 74 | |
JU476785.1 | - | 7 - 8 | 1066 | 74 | ||
JU476786.1 | - | 8 - 9 | 1114 | 74 | ||
JU476787.1 | - | 8 - 9 | 1126 | 74 | ||
JV554568.1 | - | 6 - 7 | 470 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
