Assay Details
Gene Symbol: PUS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028830187.1 | XP_028686020.1 | 1786 | 125 | ||
XM_028830188.1 | XP_028686021.1 | 1242 | 125 | |||
GenBank mRNA | JU325726.1 | - | 4 - 5 | 1081 | 125 | |
JU473311.1 | - | 5 - 6 | 1713 | 125 | ||
JV044731.1 | - | 5 - 6 | 1709 | 125 | ||
JV669348.1 | - | 5 - 6 | 1751 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
