Assay Details
Gene Symbol: FYB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015140486.2 | XP_014995972.2 | 2396 | 84 | ||
XM_028833348.1 | XP_028689181.1 | 2475 | 84 | |||
XM_028833349.1 | XP_028689182.1 | 2537 | 84 | |||
XM_028833358.1 | XP_028689191.1 | 2421 | 84 | |||
XM_028833362.1 | XP_028689195.1 | 2310 | 84 | |||
GenBank mRNA | JU481983.1 | - | 3 - 4 | 195 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
