Assay Details
Gene Symbol: FYB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015140486.2 | XP_014995972.2 | 1732 | 151 | ||
XM_028833358.1 | XP_028689191.1 | 1733 | 151 | |||
XM_028833362.1 | XP_028689195.1 | 1646 | 151 |

More Information
Set Membership: |
![]() ![]() |
