Assay Details
Gene Symbol: FYB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015140486.2 | XP_014995972.2 | 1889 | 137 | ||
XM_028833349.1 | XP_028689182.1 | 2030 | 137 | |||
XM_028833362.1 | XP_028689195.1 | 1803 | 137 |

More Information
Set Membership: |
![]() ![]() |
