Assay Details
Gene Symbol: HAO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015149229.2 | XP_015004715.1 | 996 | 65 | ||
XM_015149239.2 | XP_015004725.1 | 906 | 65 | |||
XM_015149243.2 | XP_015004729.1 | 869 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
