Assay Details
Gene Symbol: IGSF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257906.1 | NP_001244835.1 | 1 - 2 | 95 | ||
XM_028843355.1 | XP_028699188.1 | 449 | 95 | |||
GenBank mRNA | JU322970.1 | - | 1 - 2 | 95 | ||
JV044976.1 | - | 1 - 2 | 95 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001257906.1, XM_028843355.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
