Assay Details
Gene Symbol: ATF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266010.1 | NP_001252939.1 | 2 - 3 | 318 | 117 | |
XM_015116707.2 | XP_014972193.1 | 397 | 117 | |||
XM_028844193.1 | XP_028700026.1 | 4570 | 117 | |||
GenBank mRNA | JU322209.1 | - | 2 - 3 | 318 | 117 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
