Assay Details
Gene Symbol: LYPD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001107463.4 | XP_001107463.2 | 402 | 118 | ||
XM_028839319.1 | XP_028695152.1 | 502 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() |
