Assay Details
Gene Symbol: AGBL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015111938.2 | XP_014967424.1 | 2704 | 149 | ||
XM_015111940.2 | XP_014967426.1 | 2627 | 149 | |||
XM_015111941.2 | XP_014967427.1 | 3165 | 149 | |||
GenBank mRNA | JU476680.1 | - | 14 - 15 | 2588 | 149 | |
JV047854.1 | - | 14 - 15 | 2660 | 149 | ||
JV636307.1 | - | 14 - 15 | 2651 | 149 |
Gene Symbol: OST4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU325089.1 | - | 1 - 2 | 66 | 149 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
