Assay Details
Gene Symbol: CDK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028836815.1 | XP_028692648.1 | 1686 | 150 | ||
XM_028836816.1 | XP_028692649.1 | 1838 | 150 | |||
XM_028836817.1 | XP_028692650.1 | 1564 | 150 | |||
XM_028836818.1 | XP_028692651.1 | 1469 | 150 | |||
XM_028836819.1 | XP_028692652.1 | 1671 | 150 | |||
XM_028836822.1 | XP_028692655.1 | 1790 | 150 | |||
XM_028836823.1 | XP_028692656.1 | 1746 | 150 | |||
XM_028836824.1 | XP_028692657.1 | 1676 | 150 | |||
XM_028836825.1 | XP_028692658.1 | 1489 | 150 | |||
XM_028836826.1 | XP_028692659.1 | 1151 | 150 | |||
GenBank mRNA | JV637311.1 | - | 6 - 7 | 801 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
