Assay Details
Gene Symbol: GADD45A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261305.1 | NP_001248234.1 | 1 - 2 | 268 | 111 | |
GenBank mRNA | JU474506.1 | - | 1 - 2 | 268 | 111 | |
JV643929.1 | - | 1 - 2 | 321 | 111 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001261305.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
