Assay Details
Gene Symbol: FGF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001100747.3 | XP_001100747.2 | 763 | 156 |

More Information
Set Membership: |
![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FGF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001100747.3 | XP_001100747.2 | 763 | 156 |
Set Membership: |
![]() ![]() ![]() |