Assay Details
Gene Symbol: CBX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028836881.1 | XP_028692714.1 | 364 | 64 | ||
GenBank mRNA | JU477570.1 | - | 3 - 4 | 226 | 64 | |
JV664050.1 | - | 3 - 4 | 189 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
