Assay Details
Gene Symbol: OGG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260579.1 | NP_001247508.1 | 17 - 18 | 2904 | 148 |
Gene Symbol: CAMK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257871.1 | NP_001244800.1 | 1 - 2 | 112 | 148 | |
XM_015130262.2 | XP_014985748.2 | 177 | 148 | |||
XM_015130263.2 | XP_014985749.2 | 181 | 148 | |||
GenBank mRNA | JU323226.1 | - | 17 - 18 | 2904 | 148 | |
JU336824.1 | - | 1 - 2 | 112 | 148 | ||
JV699453.1 | - | 1 - 2 | 146 | 148 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001257871.1, XM_015130262.2, XM_015130263.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
