Assay Details
Gene Symbol: TNFSF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205136.3 | NP_001192065.2 | 1137 | 73 | ||
XM_015118488.2 | XP_014973974.1 | 676 | 73 | |||
XM_015118489.2 | XP_014973975.1 | 1137 | 73 | |||
XM_015118491.2 | XP_014973977.1 | 443 | 73 | |||
GenBank mRNA | JU336905.1 | - | 440 | 73 | ||
JV597155.1 | - | 697 | 73 | |||
JV668527.1 | - | 724 | 73 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() |
