Assay Details
Gene Symbol: MFHAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001090936.4 | XP_001090936.3 | 3674 | 125 | ||
GenBank mRNA | JU477842.1 | - | 2 - 3 | 3167 | 125 | |
JV635419.1 | - | 2 - 3 | 3231 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
