Assay Details
Gene Symbol: BAG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266057.2 | NP_001252986.1 | 2 - 3 | 805 | 149 | |
XM_015148170.2 | XP_015003656.2 | 822 | 149 | |||
GenBank mRNA | JU337213.1 | - | 2 - 3 | 522 | 149 | |
JU477927.1 | - | 2 - 3 | 818 | 149 | ||
JV048770.1 | - | 2 - 3 | 809 | 149 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
