Assay Details
Gene Symbol: BAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260703.1 | NP_001247632.1 | 1 - 2 | 144 | 110 | |
GenBank mRNA | JU321660.1 | - | 1 - 2 | 164 | 110 | |
JV048771.1 | - | 1 - 2 | 144 | 110 | ||
JV599021.1 | - | 1 - 2 | 185 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
