Assay Details
Gene Symbol: C1QTNF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001085902.4 | XP_001085902.2 | 150 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001085902.4 transcripts.
Set Membership: |
![]() ![]() |
