Assay Details
Gene Symbol: C1QTNF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001099341.3 | XP_001099341.2 | 908 | 81 | ||
XM_001099442.4 | XP_001099442.2 | 919 | 81 | |||
XM_028848359.1 | XP_028704192.1 | 657 | 81 | |||
GenBank mRNA | JV045661.1 | - | 2 - 3 | 276 | 81 | |
JV637266.1 | - | 2 - 3 | 375 | 81 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
