Assay Details
Gene Symbol: PLPP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001092746.4 | XP_001092746.3 | 494 | 94 | ||
XM_015145198.2 | XP_015000684.1 | 715 | 94 | |||
XM_015145199.2 | XP_015000685.1 | 441 | 94 | |||
XM_015145200.2 | XP_015000686.1 | 438 | 94 | |||
XM_015145201.2 | XP_015000687.1 | 511 | 94 | |||
XM_028852576.1 | XP_028708409.1 | 661 | 94 | |||
GenBank mRNA | JV725215.1 | - | 4 - 5 | 355 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
