Assay Details
Gene Symbol: BOK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU336055.1 | - | 1 - 2 | 124 | 123 | |
JV659564.1 | - | 1 - 2 | 129 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() |
