Assay Details
Gene Symbol: ZFP91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015114142.2 | XP_014969628.2 | 1123 | 109 | ||
GenBank mRNA | JU327536.1 | - | 440 | 109 | ||
JU477333.1 | - | 753 | 109 |

More Information
Set Membership: |
![]() ![]() |
