Assay Details
Gene Symbol: MAP3K20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001086798.4 | XP_001086798.4 | 533 | 134 | ||
XM_015110384.2 | XP_014965870.2 | 581 | 134 | |||
XM_015110385.2 | XP_014965871.2 | 386 | 134 | |||
GenBank mRNA | JU320852.1 | - | 3 - 4 | 389 | 134 | |
JU475983.1 | - | 4 - 5 | 473 | 134 | ||
JU475984.1 | - | 4 - 5 | 473 | 134 | ||
JU475985.1 | - | 4 - 5 | 473 | 134 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015110385.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
