Assay Details
Gene Symbol: GLI4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015146118.2 | XP_015001604.2 | 385 | 155 | ||
XM_015146119.2 | XP_015001605.2 | 388 | 155 | |||
XM_028853030.1 | XP_028708863.1 | 1881 | 155 | |||
XM_028853031.1 | XP_028708864.1 | 1881 | 155 | |||
GenBank mRNA | JV578700.1 | - | 3 - 4 | 295 | 155 | |
JV712304.1 | - | 3 - 4 | 308 | 155 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
