Assay Details
Gene Symbol: ROR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015141249.2 | XP_014996735.1 | 1805 | 103 | ||
XM_015141263.2 | XP_014996749.1 | 4095 | 103 | |||
GenBank mRNA | JU487395.1 | - | 1 - 2 | 56 | 103 |
Gene Symbol: LOC114669845 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028842556.1 | XP_028698389.1 | 208 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
