Assay Details
Gene Symbol: GAS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257624.1 | NP_001244553.1 | 1 - 2 | 152 | 86 | |
XM_015114619.2 | XP_014970105.1 | 539 | 86 | |||
XM_028832580.1 | XP_028688413.1 | 545 | 86 | |||
XM_028832581.1 | XP_028688414.1 | 542 | 86 | |||
XM_028832582.1 | XP_028688415.1 | 541 | 86 | |||
XM_028832583.1 | XP_028688416.1 | 539 | 86 | |||
GenBank mRNA | JU321049.1 | - | 1 - 2 | 152 | 86 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
