Assay Details
Gene Symbol: TRIM22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113359.1 | NP_001106830.1 | 3 - 4 | 747 | 112 | |
XM_015115035.2 | XP_014970521.1 | 1060 | 112 | |||
GenBank mRNA | EU124697.1 | - | 3 - 4 | 747 | 112 | |
JU332480.1 | - | 4 - 5 | 892 | 112 | ||
JU471017.1 | - | 4 - 5 | 898 | 112 | ||
JV045892.1 | - | 4 - 5 | 910 | 112 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
