Assay Details
Gene Symbol: PRG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001103106.4 | XP_001103106.2 | 622 | 72 | ||
GenBank mRNA | JU478358.1 | - | 3 - 4 | 533 | 72 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
