Assay Details
Gene Symbol: EBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015127363.2 | XP_014982849.1 | 2046 | 172 | ||
XM_015127365.1 | XP_014982851.1 | 1320 | 172 | |||
XM_028842085.1 | XP_028697918.1 | 679 | 172 | |||
XM_028842086.1 | XP_028697919.1 | 1846 | 172 | |||
GenBank mRNA | JU322615.1 | - | 3 - 4 | 531 | 172 | |
JV044645.1 | - | 3 - 4 | 533 | 172 | ||
JV531252.1 | - | 3 - 4 | 541 | 172 |
Gene Symbol: MFSD14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU471413.1 | - | 2464 | 172 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
