Assay Details
Gene Symbol: PPP1R17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193720.1 | NP_001180649.1 | 3 - 4 | 869 | 98 | |
XM_028844786.1 | XP_028700619.1 | 319 | 98 | |||
GenBank mRNA | JU322484.1 | - | 3 - 4 | 365 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
